siRNA Details
Gene Symbol: Mtag2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC107317.1 | - | 1 | 106 | ||
BC107318.1 | - | 1 | 106 | |||
BC119507.1 | - | 1 | 106 | |||
BC119508.1 | - | 1 | 106 | |||
BC127168.1 | - | 1 | 106 | |||
BC127169.1 | - | 1 | 106 | |||
U88401.1 | - | 1 | 106 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
