siRNA Details
Gene Symbol: Pdcd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016688.2 | NP_057897.2 | 3 | 1194 | ||
GenBank mRNA | AF083929.1 | - | 3 | 1188 | ||
AK155050.1 | - | 3 | 1159 | |||
BC022772.1 | - | 2 | 231 | |||
DV656988.1 | - | 2 | 207 |

More Information
Other Species Targeted

Gene Ontology Categories:
