siRNA Details
Gene Symbol: Ppp2r1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016891.3 | NP_058587.1 | 12 | 1452 | ||
GenBank mRNA | AB021743.1 | - | 12 | 1400 | ||
AK054239.1 | - | 12 | 1412 | |||
AK078135.1 | - | 12 | 1431 | |||
AK088776.1 | - | 12 | 1424 | |||
AK180835.1 | - | 3 | 143 | |||
AK186399.1 | - | 3 | 143 | |||
BC006606.1 | - | 12 | 1452 | 3157293 | ||
BC052678.1 | - | 12 | 1448 | 30062580 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
