siRNA Details
Gene Symbol: Ctps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016748.2 | NP_058028.2 | 9 | 1084 | ||
GenBank mRNA | AK146476.1 | - | 9 | 1085 | ||
AK148259.1 | - | 9 | 1033 | |||
AK161110.1 | - | 9 | 1030 | |||
BC006698.1 | - | 9 | 1059 | 3498717 | ||
U49350.1 | - | 9 | 1057 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
