siRNA Details
Gene Symbol: Rundc3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252347.1 | NP_001239276.1 | 7 | 1057 | ||
NM_016759.4 | NP_058039.1 | 7 | 1057 | |||
GenBank mRNA | AK002856.1 | - | 3 | 355 | ||
BC022745.1 | - | 5 | 782 | |||
BC046319.1 | - | 7 | 957 | 6391042 | ||
U73941.1 | - | 7 | 784 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
