siRNA Details
Gene Symbol: Ramp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016894.3 | NP_058590.1 | 3 | 1630 | ||
GenBank mRNA | AK020282.1 | - | 1 | 304 | ||
AK165238.1 | - | 3 | 1406 | |||
BC025570.1 | - | 1 | 336 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
