siRNA Details
Gene Symbol: Ramp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016894.3 | NP_058590.1 | 3 | 1004 | ||
GenBank mRNA | AB019046.1 | - | 3 | 815 | ||
AF146522.1 | - | 3 | 796 | |||
AF209904.1 | - | 3 | 775 | |||
AJ250489.1 | - | 3 | 796 | |||
AK004539.1 | - | 1 | 422 | |||
AK157896.1 | - | 3 | 794 | |||
AK165238.1 | - | 3 | 780 | |||
BC012644.1 | - | 3 | 761 | 4207524 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
