siRNA Details
Gene Symbol: Rif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175238.5 | NP_780447.4 | 21 | 2360 | ||
GenBank mRNA | AY585206.1 | - | 20 | 2353 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
