siRNA Details
Gene Symbol: Knstrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026412.3 | NP_080688.2 | 3 | 432 | ||
GenBank mRNA | AK006328.1 | - | 3 | 433 | ||
AK012148.1 | - | 3 | 232 | |||
AK013184.1 | - | 3 | 180 | |||
AK028590.1 | - | 3 | 233 | |||
AK036374.1 | - | 3 | 221 | |||
BC031709.1 | - | 2 | 140 | 4021550 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
