siRNA Details
Gene Symbol: Cdk2ap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026373.4 | NP_080649.1 | 2 | 210 | ||
GenBank mRNA | AK003686.2 | - | 2 | 176 | ||
AK012121.1 | - | 2 | 213 | |||
AK018035.1 | - | 2 | 210 | |||
BC025656.1 | - | 2 | 176 | 5342719 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
