siRNA Details
Gene Symbol: Ankrd22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024204.6 | NP_077166.4 | 3 | 567 | ||
GenBank mRNA | AK012869.1 | - | 3 | 580 | ||
AK017360.1 | - | 3 | 568 | |||
AK017408.1 | - | 3 | 452 | |||
AK037242.1 | - | 3 | 450 | |||
BC003460.1 | - | 3 | 429 | 2648395 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
