siRNA Details
Gene Symbol: Ppp1r10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163818.1 | NP_001157290.1 | 3 | 683 | ||
GenBank mRNA | AJ504718.1 | - | 3 | 689 | ||
AK053183.1 | - | 4 | 702 | |||
AK145485.1 | - | 2 | 127 | |||
BC030903.1 | - | 3 | 690 | |||
BC052059.1 | - | 3 | 686 | 5702171 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
