siRNA Details
Gene Symbol: Tmem38b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028053.2 | NP_082329.1 | 3 | 494 | ||
GenBank mRNA | AB261159.1 | - | 3 | 430 | ||
AK005479.1 | - | 3 | 477 | |||
AK136131.1 | - | 2 | 244 | |||
AK150377.1 | - | 3 | 472 | |||
AK150892.1 | - | 3 | 472 | |||
AK153075.1 | - | 3 | 459 | |||
BC011072.1 | - | 3 | 472 | 3978742 |
