siRNA Details
Gene Symbol: Tmem222 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025667.3 | NP_079943.2 | 5 | 526 | ||
GenBank mRNA | AK017503.1 | - | 5 | 522 | ||
AK079189.1 | - | 5 | 514 | |||
AK160188.1 | - | 5 | 527 | |||
BC037609.1 | - | 5 | 487 | 5356647 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
