siRNA Details
Gene Symbol: Anapc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024213.2 | NP_077175.1 | 21 | 1689 | ||
GenBank mRNA | AK011994.1 | - | 4 | 248 | ||
AK146000.1 | - | 21 | 1686 | |||
AK148625.1 | - | 21 | 1699 | |||
AK166061.1 | - | 21 | 1812 | |||
AK168897.1 | - | 22 | 1771 | |||
BC016237.1 | - | 21 | 1672 | 4504779 | ||
BC024870.1 | - | 21 | 1689 | 4949079 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
