siRNA Details
Gene Symbol: Stbd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175096.3 | NP_780305.1 | 2 | 1008 | ||
GenBank mRNA | AK050416.1 | - | 2 | 1009 | ||
BC052666.1 | - | 2 | 969 | |||
BC118615.1 | - | 3 | 1077 | 40107322 | ||
BC118661.1 | - | 2 | 991 | 40107321 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
