siRNA Details
Gene Symbol: Zfp644 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026856.2 | NP_081132.2 | 3 | 1410 | ||
GenBank mRNA | AK053146.1 | - | 3 | 1411 | ||
AK122471.1 | - | 1 | 890 | |||
AK144865.1 | - | 6 | 1586 | |||
BC057405.1 | - | 3 | 1289 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
