siRNA Details
Gene Symbol: Carhsp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025821.2 | NP_080097.1 | 4 | 1413 | ||
GenBank mRNA | AK004711.2 | - | 4 | 1413 | ||
AK143612.1 | - | 4 | 1414 | |||
AK149875.1 | - | 4 | 1417 | |||
AK150245.1 | - | 4 | 1394 | |||
AK155318.1 | - | 4 | 1416 | |||
AK158112.1 | - | 4 | 1417 | |||
AK158252.1 | - | 4 | 1416 | |||
AK165373.1 | - | 4 | 1416 | |||
AK168504.1 | - | 4 | 1414 | |||
AK170411.1 | - | 4 | 1411 | |||
AK171282.1 | - | 4 | 1416 | |||
AK210680.1 | - | 1 | 41 | |||
BC011225.1 | - | 3 | 1383 | 4166353 | ||
BC086754.1 | - | 4 | 1396 | 6812962 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
