siRNA Details
Gene Symbol: Tspan14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145928.1 | NP_666040.1 | 5 | 480 | ||
GenBank mRNA | AK030593.1 | - | 5 | 482 | ||
AK156671.1 | - | 5 | 493 | |||
BC024611.1 | - | 5 | 485 | 4188261 | ||
BC025568.1 | - | 5 | 494 | 5322531 | ||
BC026574.1 | - | 5 | 480 | 4954874 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
