siRNA Details
Gene Symbol: Ncald | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170866.1 | NP_001164337.1 | 5 | 353 | ||
NM_001170867.1 | NP_001164338.1 | 3 | 287 | |||
NM_001170868.1 | NP_001164339.1 | 2 | 235 | |||
NM_134094.4 | NP_598855.2 | 5 | 543 | |||
GenBank mRNA | AI893360.1 | - | 5 | 349 | ||
AK034031.1 | - | 3 | 266 | |||
AK048440.1 | - | 5 | 523 | |||
AK167313.1 | - | 2 | 205 | |||
BC011162.1 | - | 2 | 230 | 4191195 | ||
BC026979.1 | - | 2 | 189 | 5365560 | ||
BY258456.1 | - | 5 | 356 |

More Information
Other Species Targeted

Gene Ontology Categories:
