siRNA Details
Gene Symbol: Cbx7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144811.3 | NP_659060.1 | 1, 2 | 203 | ||
GenBank mRNA | AB221539.1 | - | 1, 2 | 77 | ||
AK045714.1 | - | 1, 2 | 197 | |||
AY453793.1 | - | 1, 2 | 193 | |||
BC021398.1 | - | 1, 2 | 219 | 5066655 | ||
BC042657.1 | - | 1, 2 | 325 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
