siRNA Details
Gene Symbol: Adgre4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139138.3 | NP_631877.2 | 3 | 230 | ||
GenBank mRNA | AF396935.1 | - | 3 | 188 | ||
AK036459.1 | - | 3 | 231 | |||
AK037483.1 | - | 3 | 175 | |||
AK155935.1 | - | 3 | 178 | |||
AY032690.1 | - | 3 | 149 | |||
BC120538.1 | - | 3 | 187 | 40129461 | ||
BC120540.1 | - | 3 | 187 | 40129463 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
