siRNA Details
Gene Symbol: Slc52a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029643.3 | NP_083919.1 | 3 | 921 | ||
GenBank mRNA | AK008081.1 | - | 3 | 900 | ||
AK029579.1 | - | 3 | 922 | |||
AK032015.1 | - | 3 | 901 | |||
AK080449.1 | - | 3 | 995 | |||
AK142597.1 | - | 3 | 1003 | |||
AK152335.1 | - | 3 | 927 | |||
AK161447.1 | - | 3 | 1033 | |||
BC016264.1 | - | 3 | 981 | 4921900 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
