siRNA Details
Gene Symbol: Pnrc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026383.3 | NP_080659.1 | 3 | 511 | ||
GenBank mRNA | AK011192.1 | - | 3 | 489 | ||
AK012073.1 | - | 3 | 495 | |||
AK013163.1 | - | 3 | 495 | |||
AK018384.1 | - | 3 | 459 | |||
AK031048.1 | - | 1509 | ||||
AK075816.1 | - | 3 | 495 | |||
AK077403.1 | - | 3 | 376 | |||
AK207137.1 | - | 1 | 107 | |||
AK210203.1 | - | 1 | 107 | |||
AK215675.1 | - | 1 | 107 | |||
AK218125.1 | - | 2 | 350 | |||
BC006598.1 | - | 3 | 484 | 3965195 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
