siRNA Details
Gene Symbol: Cnot11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028043.2 | NP_082319.1 | 7 | 1396 | ||
GenBank mRNA | AK010503.1 | - | 7 | 1408 | ||
AK041342.1 | - | 5 | 721 | |||
AK144692.1 | - | 7 | 1411 | |||
AK162118.1 | - | 1702 | ||||
AK167440.1 | - | 7 | 1387 | |||
BC049091.1 | - | 7 | 1396 | 6401756 | ||
BC050812.1 | - | 7 | 1376 | 6773595 |

More Information
Other Species Targeted

Gene Ontology Categories:
