siRNA Details
Gene Symbol: Sult1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016771.3 | NP_058051.3 | 3 | 462 | ||
GenBank mRNA | AF026073.1 | - | 3 | 340 | ||
AI118913.1 | - | 3 | 474 | |||
AK133530.1 | - | 3 | 381 | |||
AK160466.1 | - | 3 | 447 | |||
BC010752.1 | - | 3 | 435 | |||
BC066190.1 | - | 3 | 415 | 30472198 | ||
U32371.1 | - | 3 | 412 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
