siRNA Details
Gene Symbol: Banp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110100.2 | NP_001103570.1 | 13 | 1739 | ||
NM_001285981.1 | NP_001272910.1 | 12 | 1631 | |||
NM_001285983.1 | NP_001272912.1 | 12 | 1622 | |||
NM_016812.4 | NP_058092.2 | 13 | 1748 | |||
GenBank mRNA | AF091234.1 | - | 12 | 1542 | ||
AF235503.1 | - | 13 | 1659 | |||
AK154088.1 | - | 12 | 1525 | |||
AK170884.1 | - | 12 | 1538 | |||
BC013339.1 | - | 5 | 478 | |||
BC021650.1 | - | 13 | 1666 | 3660187 | ||
BC022168.1 | - | 13 | 1666 | 4189797 | ||
BC062641.1 | - | 13 | 1693 | 5718330 | ||
BU703599.1 | - | 2 | 118 | |||
DV661446.1 | - | 7 | 752 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
