siRNA Details
Gene Symbol: Banp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285981.1 | NP_001272910.1 | 8 | 1294 | ||
NM_016812.4 | NP_058092.2 | 9 | 1411 | |||
GenBank mRNA | AF091234.1 | - | 8 | 1205 | ||
AF235503.1 | - | 9 | 1322 | |||
AK209622.1 | - | 2 | 176 | |||
BC013339.1 | - | 2 | 267 | |||
BC021650.1 | - | 9 | 1329 | 3660187 | ||
BC022168.1 | - | 9 | 1329 | 4189797 | ||
BC062641.1 | - | 9 | 1356 | 5718330 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
