siRNA Details
Gene Symbol: Vamp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016796.3 | NP_058076.2 | 7 | 487 | ||
GenBank mRNA | AF061516.1 | - | 6 | 459 | ||
AK002977.1 | - | 7 | 489 | |||
AK011678.1 | - | 7 | 467 | |||
AK018344.1 | - | 7 | 498 | |||
AK031151.1 | - | 7 | 489 | |||
BC138753.1 | - | 6 | 442 | 8861775 | ||
BC138754.1 | - | 6 | 442 | 8861776 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
