siRNA Details
Gene Symbol: Mtmr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016985.2 | NP_058681.1 | 6 | 748 | ||
GenBank mRNA | AF073997.1 | - | 6 | 721 | ||
BC056376.1 | - | 6 | 748 | 6849757 | ||
BC057337.1 | - | 6 | 742 | 6825916 |
