siRNA Details
Gene Symbol: Chst3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016803.3 | NP_058083.2 | 4 | 1180 | ||
GenBank mRNA | AB008937.1 | - | 3 | 799 | ||
BC055055.1 | - | 2 | 1059 | 6493234 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
