siRNA Details
Gene Symbol: Sdcbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016807.2 | NP_058087.2 | 4, 5 | 338 | ||
GenBank mRNA | AF077527.1 | - | 4, 5 | 297 | ||
AK150661.1 | - | 4, 5 | 306 | |||
AK150736.1 | - | 4, 5 | 306 | |||
AK159323.1 | - | 4, 5 | 284 | |||
AK159538.1 | - | 4, 5 | 311 | |||
AK165657.1 | - | 4, 5 | 308 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
