siRNA Details
Gene Symbol: Sdcbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098227.1 | NP_001091697.1 | 6, 7 | 686 | ||
NM_016807.2 | NP_058087.2 | 6, 7 | 683 | |||
GenBank mRNA | AF003693.1 | - | 6, 7 | 678 | ||
AF077527.1 | - | 6, 7 | 642 | |||
AK028429.1 | - | 6, 7 | 659 | |||
AK033412.1 | - | 6, 7 | 684 | |||
AK146085.1 | - | 6, 7 | 658 | |||
AK150278.1 | - | 6, 7 | 651 | |||
AK150321.1 | - | 6, 7 | 654 | |||
AK150544.1 | - | 6, 7 | 658 | |||
AK150623.1 | - | 6, 7 | 662 | |||
AK150661.1 | - | 6, 7 | 651 | |||
AK150707.1 | - | 6, 7 | 657 | |||
AK150721.1 | - | 6, 7 | 666 | |||
AK150736.1 | - | 6, 7 | 651 | |||
AK150801.1 | - | 6, 7 | 656 | |||
AK150996.1 | - | 6, 7 | 658 | |||
AK151050.1 | - | 6, 7 | 658 | |||
AK152001.1 | - | 6, 7 | 657 | |||
AK152017.1 | - | 6, 7 | 658 | |||
AK152109.1 | - | 6, 7 | 658 | |||
AK152434.1 | - | 6, 7 | 658 | |||
AK152552.1 | - | 6, 7 | 662 | |||
AK152577.1 | - | 6, 7 | 657 | |||
AK152855.1 | - | 6, 7 | 658 | |||
AK153042.1 | - | 6, 7 | 658 | |||
AK153112.1 | - | 6, 7 | 662 | |||
AK159124.1 | - | 6, 7 | 654 | |||
AK159323.1 | - | 6, 7 | 629 | |||
AK159538.1 | - | 6, 7 | 656 | |||
AK159677.1 | - | 6, 7 | 665 | |||
AK165657.1 | - | 6, 7 | 653 | |||
AK167513.1 | - | 6, 7 | 654 | |||
AK168004.1 | - | 6, 7 | 661 | |||
AK168777.1 | - | 6, 7 | 654 | |||
AK169219.1 | - | 6, 7 | 663 | |||
AK169415.1 | - | 6, 7 | 692 | |||
AK169687.1 | - | 6, 7 | 658 | |||
AK171162.1 | - | 6, 7 | 658 | |||
AK178827.1 | - | 2, 3 | 244 | |||
AK193537.1 | - | 1, 2 | 89 | |||
AK206044.1 | - | 1, 2 | 89 | |||
AK212103.1 | - | 1, 2 | 89 | |||
AK213782.1 | - | 2, 3 | 244 | |||
BC019400.1 | - | 6, 7 | 649 | 4017442 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
