siRNA Details
Gene Symbol: Psmd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164177.1 | NP_001157649.1 | 4 | 457 | ||
NM_016883.4 | NP_058579.2 | 4 | 457 | |||
GenBank mRNA | AB022022.1 | - | 4 | 413 | ||
AK009068.1 | - | 4 | 436 | |||
AK018233.1 | - | 4 | 435 | |||
AK136400.1 | - | 4 | 436 | |||
BC026931.1 | - | 4 | 424 | 3987112 | ||
BC056196.1 | - | 4 | 442 | 30019985 | ||
CD564300.1 | - | 4 | 424 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
