siRNA Details
Gene Symbol: Hif3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162950.1 | NP_001156422.1 | 9 | 1114 | ||
NM_016868.3 | NP_058564.2 | 9 | 1145 | |||
GenBank mRNA | AB289606.1 | - | 9 | 1081 | ||
AF060194.1 | - | 9 | 1145 | |||
AK144472.1 | - | 4 | 488 | |||
BC120587.1 | - | 9 | 1098 | 40129510 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
