siRNA Details
Gene Symbol: Syt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016908.2 | NP_058604.1 | 7 | 1124 | ||
GenBank mRNA | AB026806.1 | - | 7 | 868 | ||
BC047148.1 | - | 7 | 1124 | 6465481 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
