siRNA Details
Gene Symbol: Snrpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001046637.1 | NP_001040102.1 | 2 | 472 | ||
NM_001285825.1 | NP_001272754.1 | 3 | 546 | |||
NM_015782.3 | NP_056597.3 | 3 | 549 | |||
GenBank mRNA | AK013886.1 | - | 3 | 523 | ||
AK088550.1 | - | 3 | 407 | |||
BC003229.1 | - | 3 | 449 | 3483391 | ||
BC094006.1 | - | 3 | 409 | 4983854 | ||
BC096648.1 | - | 3 | 403 | 1397390 | ||
CX206945.1 | - | 3 | 549 | |||
CX565356.1 | - | 2 | 442 | |||
DV650448.1 | - | 1 | 165 | |||
L15447.1 | - | 3 | 345 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
