siRNA Details
Gene Symbol: Map3k6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016693.5 | NP_057902.5 | 16 | 2358 | ||
GenBank mRNA | AB021861.4 | - | 16 | 2356 | ||
AK204377.1 | - | 2 | 112 | |||
BC120565.1 | - | 16 | 2285 | 40129488 | ||
BC125577.1 | - | 16 | 2285 | 40130092 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
