siRNA Details
Gene Symbol: Clasrp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016680.5 | NP_057889.3 | 12 | 1077 | ||
GenBank mRNA | AB080582.1 | - | 11 | 940 | ||
AB080583.1 | - | 11 | 940 | |||
AF042799.1 | - | 12 | 1042 | |||
AF325456.1 | - | 2 | 923 | |||
AK009334.1 | - | 12 | 1028 | |||
AK159443.1 | - | 12 | 1046 | |||
BC010728.1 | - | 12 | 1028 | |||
BC051916.1 | - | 12 | 1052 | 5707517 | ||
BC060092.1 | - | 11 | 3445 | |||
BC079543.1 | - | 11 | 3435 | |||
BC103781.1 | - | 10 | 907 | |||
BC106129.1 | - | 12 | 1040 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
