siRNA Details
Gene Symbol: Vamp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080742.2 | NP_001074211.1 | 3 | 254 | ||
NM_016872.4 | NP_058568.1 | 3 | 254 | |||
GenBank mRNA | AF035643.1 | - | 2 | 178 | ||
AK009266.1 | - | 3 | 257 | |||
AK145137.1 | - | 2 | 1958 | |||
BC086925.1 | - | 3 | 208 | 6758831 | ||
BY709062.1 | - | 3 | 251 |

More Information
Other Species Targeted

Gene Ontology Categories:
