siRNA Details
Gene Symbol: Cldn7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193619.1 | NP_001180548.1 | 4 | 614 | ||
NM_016887.6 | NP_058583.1 | 3 | 936 | |||
GenBank mRNA | AF087825.1 | - | 3 | 430 | ||
AK002924.1 | - | 3 | 857 | |||
AK010558.1 | - | 3 | 856 | |||
AK087296.1 | - | 3 | 854 | |||
AK145504.1 | - | 3 | 937 | |||
AK167250.1 | - | 3 | 857 | |||
BC008104.1 | - | 3 | 501 | 3157774 | ||
BC050007.1 | - | 3 | 849 | 6530942 | ||
BG083098.2 | - | 1 | 57 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
