siRNA Details
Gene Symbol: Zranb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017381.2 | NP_059077.1 | 1 | 68 | ||
GenBank mRNA | AF133818.1 | - | 1 | 46 | ||
AK146275.1 | - | 1 | 66 | |||
AK146854.1 | - | 1 | 60 | |||
AK150291.1 | - | 1 | 107 | |||
AK152536.1 | - | 1 | 107 | |||
AK160862.1 | - | 1 | 65 | |||
AK166534.1 | - | 1 | 63 | |||
AK167756.1 | - | 1 | 66 | |||
AK167766.1 | - | 1 | 96 | |||
AK167862.1 | - | 1 | 96 | |||
AK167983.1 | - | 1 | 96 | |||
AK168033.1 | - | 1 | 66 | |||
AK168162.1 | - | 1 | 67 | |||
AK168613.1 | - | 1 | 98 | |||
AK169114.1 | - | 1 | 60 | |||
AK169335.1 | - | 1 | 69 | |||
BC049115.1 | - | 1 | 78 | |||
BC132547.1 | - | 1 | 65 | 40130824 | ||
BC138575.1 | - | 1 | 65 | 8861595 |

More Information
Other Species Targeted

Gene Ontology Categories:
