siRNA Details
Gene Symbol: Zranb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017381.2 | NP_059077.1 | 7 | 593 | ||
GenBank mRNA | AF133818.1 | - | 7 | 571 | ||
AK146275.1 | - | 7 | 591 | |||
AK146854.1 | - | 7 | 585 | |||
AK150291.1 | - | 7 | 632 | |||
AK152536.1 | - | 7 | 632 | |||
AK160862.1 | - | 6 | 481 | |||
AK166534.1 | - | 7 | 588 | |||
AK167756.1 | - | 7 | 591 | |||
AK167766.1 | - | 7 | 621 | |||
AK167862.1 | - | 7 | 621 | |||
AK167983.1 | - | 7 | 621 | |||
AK168033.1 | - | 7 | 591 | |||
AK168162.1 | - | 7 | 592 | |||
AK168613.1 | - | 7 | 623 | |||
AK169114.1 | - | 7 | 585 | |||
AK169335.1 | - | 7 | 594 | |||
BC049115.1 | - | 7 | 603 | |||
BC051187.1 | - | 2 | 180 | 6478334 | ||
BC132547.1 | - | 7 | 590 | 40130824 | ||
BC138575.1 | - | 7 | 590 | 8861595 |
