siRNA Details
Gene Symbol: Nphp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291012.1 | NP_001277941.1 | 12 | 1173 | ||
NM_001291013.1 | NP_001277942.1 | 12 | 1170 | |||
NM_016902.4 | NP_058598.1 | 12 | 1176 | |||
GenBank mRNA | AF127180.1 | - | 12 | 1127 | ||
AF176210.1 | - | 12 | 1139 | |||
AJ243223.1 | - | 12 | 1124 | |||
AK159627.1 | - | 12 | 1150 | |||
BC118953.2 | - | 12 | 1121 | 40093235 | ||
EF174553.3 | - | 12 | 1145 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
