siRNA Details
Gene Symbol: Sart3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016926.1 | NP_058622.1 | 7 | 1069 | ||
GenBank mRNA | AF172722.1 | - | 7 | 1069 | ||
AK053828.1 | - | 7 | 1073 | |||
AK086398.1 | - | 7 | 1070 | |||
AK129067.1 | - | 7 | 1064 | |||
BC036350.1 | - | 7 | 1070 | |||
BC057156.1 | - | 7 | 1060 | 5347469 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
