siRNA Details
Gene Symbol: Rcan2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286653.1 | NP_001273582.1 | 3 | 399 | ||
NM_001286654.1 | NP_001273583.1 | 3 | 276 | |||
NM_030598.2 | NP_085101.1 | 2 | 406 | |||
NM_207649.1 | NP_997532.1 | 3 | 532 | |||
GenBank mRNA | AB061524.1 | - | 2 | 344 | ||
AB061525.1 | - | 3 | 399 | |||
AF237791.1 | - | 2 | 146 | |||
AF237887.1 | - | 2 | 192 | |||
AK049006.1 | - | 2 | 345 | |||
AK136805.1 | - | 3 | 340 | |||
AK138768.1 | - | 3 | 444 | |||
AK160865.1 | - | 2 | 294 | |||
BB635249.1 | - | 3 | 444 | |||
BC047153.1 | - | 2 | 204 | 6491318 | ||
BC049096.1 | - | 2 | 406 | 6416901 | ||
BC062141.1 | - | 3 | 276 | 30290791 | ||
BF534085.1 | - | 379 | ||||
BY740155.1 | - | 3 | 539 |

More Information
Other Species Targeted

Gene Ontology Categories:
