siRNA Details
Gene Symbol: Rps28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC010987.1 | - | 4 | 278 | 4218716 | |
BC090982.1 | - | 4 | 276 | 30316901 | ||
DQ566373.1 | - | 12 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
