siRNA Details
Gene Symbol: Acrbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016845.2 | NP_058541.2 | 6 | 1061 | ||
GenBank mRNA | AK132705.1 | - | 6 | 1029 | ||
D17573.1 | - | 6 | 1013 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
