siRNA Details
Gene Symbol: Avpr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016847.2 | NP_058543.2 | 1 | 1282 | ||
GenBank mRNA | AK137203.1 | - | 1 | 1210 | ||
AK149578.1 | - | 1 | 1225 | |||
AK155952.1 | - | 1 | 1215 | |||
BC024149.1 | - | 1 | 1280 | 4220075 | ||
D49729.1 | - | 1 | 1264 | |||
D49730.1 | - | 1 | 1288 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
