siRNA Details
Gene Symbol: Rdh7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001150749.1 | NP_001144221.1 | 3 | 830 | ||
NM_017473.4 | NP_059501.1 | 3 | 802 | |||
GenBank mRNA | AB015165.1 | - | 3 | 918 | ||
AB032058.1 | - | 3 | 710 | |||
AF056194.1 | - | 3 | 807 | |||
BC024603.1 | - | 3 | 780 | 4196075 | ||
BC092255.1 | - | 3 | 756 | 5100891 | ||
BC093514.1 | - | 3 | 744 | 5132290 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
